Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

311316

Sigma-Aldrich

4-Methyl-5-thiazolylethyl acetate

99%

Synonym(s):

Sulfurol acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11NO2S
CAS Number:
Molecular Weight:
185.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

99%

form

liquid

refractive index

n20/D 1.51 (lit.)

bp

117-118 °C/6 mmHg (lit.)

density

1.147 g/mL at 25 °C (lit.)

SMILES string

CC(=O)OCCc1scnc1C

InChI

1S/C8H11NO2S/c1-6-8(12-5-9-6)3-4-11-7(2)10/h5H,3-4H2,1-2H3

InChI key

CRTCWNPLKVVXIX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service