Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

311286

Sigma-Aldrich

Nonyl phenyl ketone

≥99%

Synonym(s):

Decanophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CO(CH2)8CH3
CAS Number:
Molecular Weight:
232.36
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥99%

form

solid

bp

168 °C/5 mmHg (lit.)

mp

34-36 °C (lit.)

functional group

ketone
phenyl

SMILES string

CCCCCCCCCC(=O)c1ccccc1

InChI

1S/C16H24O/c1-2-3-4-5-6-7-11-14-16(17)15-12-9-8-10-13-15/h8-10,12-13H,2-7,11,14H2,1H3

InChI key

QQXJNLYVPPBERR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nonyl phenyl ketone (Decanophenone) serves as the best micelle marker to date. It has a strong chromophore which is detectable at all pH levels and is easy to dissolve in the mobile phase[1].

Application

HPLC standard

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

D J Bailey et al.
Journal of chromatography. A, 852(2), 559-571 (1999-09-11)
In many biological and environmental situations, the pH of aqueous media varies and differences in solute partitioning may result. However, the majority of biopartitioning and hydrophobicity studies conducted have been at pH 7. Using migration factors measured by micellar electrokinetic
Jie Lu et al.
Journal of chromatography. A, 1359, 296-302 (2014-07-30)
In this paper, a novel pseudostationary phase (PSP), the vesicle formed from octyltriethylammonium bromide (C8NE3Br) and sodium dodecyl benzene sulfonate (SDBS), has been developed in electrokinetic chromatography (EKC). Physicochemical parameters of the mixture of catanionic surfactants such as ζ potential
Azza H Rageh et al.
Electrophoresis, 36(5), 784-795 (2014-09-27)
In this work, we extend our investigations regarding the separation of urinary nucleosides by MEKC with the ionic liquid type surfactant 1-tetradecyl-3-methylimidazolium bromide (C14MImBr). We study the impact of adding alkyl- and arylboronic acids (in the presence of C14MImBr micelles)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service