Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

310689

Sigma-Aldrich

Tetramethylammonium borohydride

95%

Synonym(s):

NSC 164901, TMAB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)4N(BH4)
CAS Number:
Molecular Weight:
88.99
Beilstein/REAXYS Number:
3684968
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

powder

reaction suitability

reagent type: reductant

functional group

amine

SMILES string

[BH4-].C[N+](C)(C)C

InChI

1S/C4H12N.BH4/c1-5(2,3)4;/h1-4H3;1H4/q+1;-1

InChI key

LSSMEJCYZMCNIN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Preparation of iridium tetrahydride complexes bearing a tridentate pincer-type bis(phosphino)silyl ligand
  • Reduction reactions (reducing agent)

pictograms

FlameSkull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Water-react 2

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isabelle Russier-Antoine et al.
Nanoscale, 8(5), 2892-2898 (2016-01-15)
We demonstrate that the tuning of the light emission from red to blue in dihydrolipoic acid (DHLA) capped Ag29 nanoclusters can be trigged with one and two photon excitations. The cluster stoichiometry was determined with mass spectrometry and found to
Hilton B Swan et al.
Analytical and bioanalytical chemistry, 409(18), 4409-4423 (2017-05-21)
Dimethylsulfoniopropionate (DMSP) and eleven other target zwitterions were quantified in the branch tips of six Acropora species and Stylophora pistillata hard coral growing on the reef flat surrounding Heron Island in the southern Great Barrier Reef (GBR), Australia. Hydrophilic interaction
Abdulatef Ahhmed et al.
Meat science, 127, 35-44 (2017-01-24)
The effects of utilizing fermented soybeans paste (miso) as an alternative flavor-coating material for eliminating unpleasant odor of sulphuric and sotolone compounds from cemen in commercial pastirma were investigated. Results showed that miso-pastirma (MP) and commercial pastirma (CP) had higher
Ronnie Mooney et al.
Scientific reports, 10(1), 6420-6420 (2020-04-15)
The opportunistic pathogen, Acanthamoeba castellanii is the causative agent for the sight threatening infection Acanthamoeba keratitis (AK). It is commonly associated with contact lens wearers, and prevalence is increasing at an alarming rate due to an inadequate preventive strategy to
Volker Maus et al.
Cerebrovascular diseases (Basel, Switzerland), 44(3-4), 113-121 (2017-06-13)
Cerebral large vessel occlusion (LVO) in acute ischemic stroke (AIS) may be complete (CLVO) or incomplete (ILVO). The influence of ILVO on clinical outcome after mechanical thrombectomy (MT) remains unclear. We investigated primarily the clinical outcome in patients with AIS

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service