About This Item
Recommended Products
assay
97%
refractive index
n20/D 1.464 (lit.)
bp
98-99 °C/1 mmHg (lit.)
density
1.014 g/mL at 25 °C (lit.)
functional group
ester
nitrile
SMILES string
CCOC(=O)\C(C#N)=C(\C)C
InChI
1S/C8H11NO2/c1-4-11-8(10)7(5-9)6(2)3/h4H2,1-3H3
InChI key
PZMDAADKKAXROL-UHFFFAOYSA-N
General description
signalword
Warning
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
224.6 °F - closed cup
flash_point_c
107 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service