Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

310395

Sigma-Aldrich

(1-Hydroxyallyl)trimethylsilane

97%

Synonym(s):

1-(Trimethylsilyl)-2-propen-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14OSi
CAS Number:
Molecular Weight:
130.26
MDL number:
Pricing and availability is not currently available.

assay

97%

SMILES string

C[Si](C)(C)C(O)C=C

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

98.6 °F - closed cup

flash_point_c

37 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service