Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

310174

Sigma-Aldrich

Sodium hydrogen difluoride

pellets, diam. × L ~5 mm × 5 mm, 99%

Synonym(s):

Sodium acid fluoride, Sodium bifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaHF2
CAS Number:
Molecular Weight:
61.99
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
99%
grade:
for analytical purposes
form:
pellets

grade

for analytical purposes

assay

99%

form

pellets

diam. × L

~5 mm × 5 mm

mp

>68 °C (dec.)

SMILES string

[F-].[Na+].F[H]

InChI

1S/2FH.Na/h2*1H;/q;;+1/p-1

InChI key

BFXAWOHHDUIALU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Starting material for solid state preparation of Na3MnF6 for further high pressure structural determination studies.[1]

pictograms

Skull and crossbonesCorrosion

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carlson, S. et al.
Inorganic Chemistry, 37, 1486-1486 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service