309524
Poly(ethylene glycol) tetrahydrofurfuryl ether
average Mn 200
Synonym(s):
Mixture of diethylene and triethylene glycol mono-tetrahydrofurfuryl ether, Tetrahydrofurfuryl polyethylene glycol 200
About This Item
Recommended Products
mol wt
average Mn 200
refractive index
n20/D 1.462 (lit.)
density
1.069 g/mL at 25 °C (lit.)
Ω-end
hydroxyl
α-end
tetrahydrofurfuryl ether
InChI
1S/C7H14O3/c8-3-5-9-6-7-2-1-4-10-7/h7-8H,1-6H2
InChI key
CTPDSKVQLSDPLC-UHFFFAOYSA-N
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
275.0 °F - closed cup
flash_point_c
135 °C - closed cup
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Progress in biotechnology fields such as tissue engineering and drug delivery is accompanied by an increasing demand for diverse functional biomaterials. One class of biomaterials that has been the subject of intense research interest is hydrogels, because they closely mimic the natural environment of cells, both chemically and physically and therefore can be used as support to grow cells. This article specifically discusses poly(ethylene glycol) (PEG) hydrogels, which are good for biological applications because they do not generally elicit an immune response. PEGs offer a readily available, easy to modify polymer for widespread use in hydrogel fabrication, including 2D and 3D scaffold for tissue culture. The degradable linkages also enable a variety of applications for release of therapeutic agents.
Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service