Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

307602

Sigma-Aldrich

Ammonium hydrogensulfate

98%

Synonym(s):

Ammonium bisulfate, Ammonium sulfate monobasic

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)HSO4
CAS Number:
Molecular Weight:
115.11
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

98%

bp

350 °C (dec.)(lit.)

mp

121-145 °C (lit.)

density

1.79 g/mL at 25 °C (lit.)

SMILES string

N.OS(O)(=O)=O

InChI

1S/H3N.H2O4S/c;1-5(2,3)4/h1H3;(H2,1,2,3,4)

InChI key

BIGPRXCJEDHCLP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ammonium bisulfite contact dermatitis: face eczema due to a bleaching ointment used during hair-dying.
A Nassif
Contact dermatitis, 55(2), 124-124 (2006-08-26)
M J Utell
Environmental health perspectives, 63, 39-44 (1985-11-01)
There exist significant gaps in our understanding of human health effects from inhalation of pollutants associated with acid precipitation. Controlled clinical studies examine effects of criteria pollutants almost exclusively by assessing changes in lung mechanics. One constituent of acid precipitation
B G Ferris et al.
Environmental health perspectives, 63, 5-9 (1985-11-01)
Problems associated with estimation of human exposure to ambient air pollutants are discussed. Ideally, we would prefer to have some indication of actual dose. For most pollutants this is not presently feasible. Specific problems discussed are adequacy of outdoor monitors;
Bryan R Bzdek et al.
The journal of physical chemistry. A, 114(43), 11638-11644 (2010-10-13)
The reaction kinetics of ammonium bisulfate clusters with dimethylamine (DMA) gas were investigated using Fourier transform ion cyclotron resonance mass spectrometry (FTICR-MS). Clusters ranged in size from 1 to 10 bisulfate ions. Although displacement of the first several ammonium ions
W S Tunnicliffe et al.
Occupational and environmental medicine, 60(11), e15-e15 (2003-10-24)
To explore the effects in normal and asthmatic adults of exposure to 200 ppb sulphur dioxide (SO2) and 200 microg/m3 and 2000 microg/m3 aerosols of ammonium bisulphate (AB) and sulphuric acid (SA) (MMD 0.3 microm). Exposures were placebo controlled, for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service