Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

306819

Sigma-Aldrich

1,6-Diisocyanohexane

98%

Synonym(s):

Hexamethylene diisocyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CNCH2CH2CH2CH2CH2CH2NC
CAS Number:
Molecular Weight:
136.19
Beilstein/REAXYS Number:
3662465
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

98%

form

liquid

refractive index

n20/D 1.443 (lit.)

bp

101-103 °C/0.5 mmHg (lit.)

mp

21-23 °C (lit.)

density

0.899 g/mL at 25 °C (lit.)

SMILES string

[C-]#[N+]CCCCCC[N+]#[C-]

InChI

1S/C8H12N2/c1-9-7-5-3-4-6-8-10-2/h3-8H2

InChI key

WJCPMQQLTJQIJK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,6-Diisocyanohexane was used in the synthesis of unusual tricyclic macroheterocycles by reacting with 1,1-diisityl-3,3,3-triisopropyldisilaphosphene and analogous disilaarsene[1].

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Eye Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

219.2 °F - closed cup

flash_point_c

104 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel type of oligofunctional macroheterocycle by reaction of silylidene-phosphanes and-arsanes with 1, 6-diisocyanohexane.
Journal of the Chemical Society. Chemical Communications, 20, 1585-1587 (1993)
Didem Mumcuoglu et al.
Materials science & engineering. C, Materials for biological applications, 84, 271-280 (2018-03-10)
Bone morphogenetic protein-2 (BMP-2) is a powerful osteoinductive protein; however, there is a need for the development of a safe and efficient BMP-2 release system for bone regeneration therapies. Recombinant extracellular matrix proteins are promising next generation biomaterials since the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service