Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

305960

Sigma-Aldrich

2-Hydroxy-3-methoxybenzyl alcohol

97%

Synonym(s):

o-Vanillyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(OCH3)CH2OH
CAS Number:
Molecular Weight:
154.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

bp

135 °C/0.2 mmHg (lit.)

mp

61-63 °C (lit.)

SMILES string

COc1cccc(CO)c1O

InChI

1S/C8H10O3/c1-11-7-4-2-3-6(5-9)8(7)10/h2-4,9-10H,5H2,1H3

InChI key

OSZHSESNQIMXMZ-UHFFFAOYSA-N

Application

2-Hydroxy-3-methoxybenzyl alcohol was employed as substrate, to investigate in vitro substrate specificity of UDP-glucose:p-hydroxymandelonitrile-O-glucosyltransferase from Sorghum bicolor (UGT85B1).[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karina Sinding Hansen et al.
Phytochemistry, 64(1), 143-151 (2003-08-30)
The in vitro substrate specificity of UDP-glucose:p-hydroxymandelonitrile-O-glucosyltransferase from Sorghum bicolor (UGT85B1) was examined using a range of potential acceptor molecules, including cyanohydrins, terpenoids, phenolics, hexanol derivatives and plant hormones. Qualitative enzyme activity assays employing 20 different putative substrates were performed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service