Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

303887

Sigma-Aldrich

1-Methyl-2-[(phenylsulfonyl)methyl]benzene

97%

Synonym(s):

2-Methylbenzyl phenyl sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CH2SO2C6H5
CAS Number:
Molecular Weight:
246.32
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

form

solid

mp

81-83 °C (lit.)

SMILES string

Cc1ccccc1CS(=O)(=O)c2ccccc2

InChI

1S/C14H14O2S/c1-12-7-5-6-8-13(12)11-17(15,16)14-9-3-2-4-10-14/h2-10H,11H2,1H3

InChI key

PWECXBSVNSKIIN-UHFFFAOYSA-N

Application

1-Methyl-2-[(phenylsulfonyl)methyl]benzene (2-Methylbenzyl phenyl sulfone) was used in the synthesis of nitrile derivatives.[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C-cyanation with 1-cyanobenzotriazole
Katritzky AR, et al.
ARKIVOC (Gainesville, FL, United States), 3, 5-12 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service