Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

303046

Sigma-Aldrich

2′-Hydroxy-6′-methoxyacetophenone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(OCH3)COCH3
CAS Number:
Molecular Weight:
166.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

bp

141 °C/16 mmHg (lit.)

mp

58-60 °C (lit.)

SMILES string

COc1cccc(O)c1C(C)=O

InChI

1S/C9H10O3/c1-6(10)9-7(11)4-3-5-8(9)12-2/h3-5,11H,1-2H3

InChI key

UENLHUMCIOWYQN-UHFFFAOYSA-N

General description

An organometallic synthetic route to 2′-hydroxy-6′-methoxyacetophenone was reported.[1]

Application

2′-Hydroxy-6′-methoxyacetophenone was used as starting material in the synthesis of 5-methoxyflavaone.[2] It was also used in synthesis of 5,6-dihydroxyflavone.[3]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A convenient large-scale synthesis of 5-methoxyflavone and its application to analog preparation
Ares JJ, et al.
The Journal of Organic Chemistry, 58(27), 7903-7905 (1993)
An Organometallic Route to 2'-Hydroxy-6'-methoxy-acetophenone.
Mincheva Z, et al.
Synthetic Communications, 25(2), 149-155 (1995)
204. The synthesis of 5: 6-dihydroxyflavone and the structure of primetin.
Baker W.
Journal of the Chemical Society, 956-961 (1939)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service