Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

302821

Sigma-Aldrich

1-Benzyl-3-pyrrolidinol

95%

Synonym(s):

1-Benzyl-3-hydroxypyrrolidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15NO
CAS Number:
Molecular Weight:
177.24
Beilstein/REAXYS Number:
6039
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

liquid

refractive index

n20/D 1.548 (lit.)

bp

113-115 °C/2 mmHg (lit.)

density

1.07 g/mL at 25 °C (lit.)

functional group

hydroxyl
phenyl

SMILES string

OC1CCN(C1)Cc2ccccc2

InChI

1S/C11H15NO/c13-11-6-7-12(9-11)8-10-4-2-1-3-5-10/h1-5,11,13H,6-9H2

Inchi Key

YQMXOIAIYXXXEE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Benzyl-3-pyrrolidinol was used in preparation of:
  • potent calcium antagonist, 1-benzyl-3-pyrrolidinyl methyl 2,6-dimethyl-4-(m-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate[1]
  • (3R)-1-benzyl-3-methanesulfonyloxy pyrrolidine[2]

Pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Tamazawa et al.
Journal of medicinal chemistry, 29(12), 2504-2511 (1986-12-01)
Four enantiomers (3a-d) of the title compound, YM-09730 (3), were synthesized by the reaction of (-)- or (+)-5-(methoxycarbonyl)-2, 6-dimethyl-4-(m-nitrophenyl)-1,4-dihydropyridine-3-carboxylic acid (1a or 1b) with (S)- or (R)-1-benzyl-3-pyrrolidinol (2a or 2b). [3H]Nitrendipine binding affinity and coronary vasodilating activity of these compounds
Efficient synthesis of the 5-HT2c receptor agonist, Org 37684.
Adams DR and Duncton MAJ.
Synthetic Communications, 31(13), 2029-2036 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service