Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

301957

Sigma-Aldrich

10-Camphorsulfonyl chloride

98%

Synonym(s):

Camphor-10-sulfonic acid chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15ClO3S
CAS Number:
Molecular Weight:
250.74
Beilstein/REAXYS Number:
3205975
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

powder

mp

66-68 °C (lit.)

SMILES string

CC1(C)[C@H]2CC[C@]1(CS(Cl)(=O)=O)C(=O)C2

InChI

1S/C10H15ClO3S/c1-9(2)7-3-4-10(9,8(12)5-7)6-15(11,13)14/h7H,3-6H2,1-2H3/t7-,10-/m0/s1

InChI key

BGABKEVTHIJBIW-XVKPBYJWSA-N

Application

10-Camphorsulfonyl chloride was used in preparation of 1,2-bis(hydroxycamphorsulfonamido)cyclohexenes[1] and 10-mercaptoisoborneol.[2]

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric synthesis of nearly optically pure atrolactic acid methyl ether.
Eliel EL and Frazee WJ.
The Journal of Organic Chemistry, 44(20), 3598-3599 (1979)
Highly enantioselective addition of dialkylzinc reagents to ketones promoted by titanium tetraisopropoxide.
Yus M, et al.
Tetrahedron Asymmetry, 13(21), 2291-2293 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service