Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

301469

Sigma-Aldrich

(S)-(+)-5-Oxo-2-tetrahydrofurancarboxylic acid

98%

Synonym(s):

S-Hothf, (S)-γ-Carboxy-γ-butyrolactone, (S)-5-Oxotetrahydro-2-furancarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6O4
CAS Number:
Molecular Weight:
130.10
Beilstein/REAXYS Number:
1424498
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

98%

optical activity

[α]20/D +14°, c = 5 in methanol

optical purity

ee: 99% (GLC)

bp

150-155 °C/0.2 mmHg (lit.)

mp

71-73 °C (lit.)

functional group

carboxylic acid
ester

SMILES string

OC(=O)[C@@H]1CCC(=O)O1

InChI

1S/C5H6O4/c6-4-2-1-3(9-4)5(7)8/h3H,1-2H2,(H,7,8)/t3-/m0/s1

InChI key

QVADRSWDTZDDGR-VKHMYHEASA-N

General description

(S)-(+)-5-Oxo-2-tetrahydrofurancarboxylic acid is a chiral derivatizing agent for alcohols.[1]

Application

(S)-(+)-5-Oxo-2-tetrahydrofurancarboxylic acid may be used as a ligand to form water-soluble silver(I) complexes by reacting with silver oxide. These complexes show good antibacterial and antifungal activities.[1] It may also be used a chiral resolving agent during the multi-step synthesis of phosphatidylinositol 3,5-bisphosphate [PtdIns(3,5)P2].[2]
Used to prepare municatacin analogs which show cytotoxic activity on tumor cells.[3]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Figadere, B.
Tetrahedron Letters, 32, 7539-7539 (1991)
Synthesis and crystal structure of three silver (I) complexes with (S)-(+)-5-oxo-2-tetrahydrofurancarboxylic acid (S-Hothf) and its isomeric forms (R-Hothf and R, S-Hothf) showing wide spectra of effective antibacterial and antifungal activities. ....
Nomiya K, et al.
J. Chem. Soc., Dalton Trans., 8, 1343-1348 (2000)
Hongde Li et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(6), 1353-1358 (2017-01-25)
L-2-hydroxyglutarate (L-2HG) has emerged as a putative oncometabolite that is capable of inhibiting enzymes involved in metabolism, chromatin modification, and cell differentiation. However, despite the ability of L-2HG to interfere with a broad range of cellular processes, this molecule is
l-a-Phosphatidyl-d-myo-inositol 3, 5-bisphosphate: total synthesis of a new inositol phospholipid via myo-inositol orthoacetate.
Riley AM and Potter BVL
Tetrahedron Letters, 39(37), 6769-6772 (1998)
Shinji K Strain et al.
Rapid communications in mass spectrometry : RCM, 33(17), 1401-1409 (2019-05-31)
2-Hydroxyglutarate (2-hg) exists as enantiomers and can readily undergo cyclization to its lactone. Gas chromatography/electron ionization mass spectrometry (GC/EI-MS) has been used to separate 2-hg enantiomers in bodily fluids but the assay cannot simultaneously measure cyclic and acylic 2-hg enantiomers.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service