Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

301256

Sigma-Aldrich

Trimethylamine

anhydrous, 99%

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103

grade

anhydrous

assay

99%

form

gas

Packaging

Packaged in steel cylinders each fitted with stainless steel valve

Recommended products

Stainless steel regulator Z148504 or Z148512 is recommended.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service