Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

295957

Sigma-Aldrich

(S)-(+)-1-(1-Naphthyl)ethyl isocyanate

99%

Synonym(s):

(S)-NEI

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7CH(CH3)NCO
CAS Number:
Molecular Weight:
197.23
Beilstein/REAXYS Number:
4675071
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

assay

99%

optical activity

[α]20/D +45°, c = 3.5 in toluene

optical purity

ee: 96% (GLC)

quality

stabilized

refractive index

n20/D 1.604 (lit.)

bp

106-108 °C/0.16 mmHg (lit.)

density

1.128 g/mL at 25 °C (lit.)

Storage temp.

2-8°C

SMILES string

C[C@H](N=C=O)c1cccc2ccccc12

InChI

1S/C13H11NO/c1-10(14-9-15)12-8-4-6-11-5-2-3-7-13(11)12/h2-8,10H,1H3/t10-/m0/s1

Inchi Key

GONOHGQPZFXJOJ-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

199.4 °F - closed cup

flash_point_c

93 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service