Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

295604

Sigma-Aldrich

Oxygen

≥99.6%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2
CAS Number:
Molecular Weight:
32.00
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:

vapor density

1.11 (vs air)

Quality Level

assay

≥99.6%

impurities

≤10 ppm water

bp

−183 °C (lit.)

mp

−218 °C (lit.)

SMILES string

O=O

InChI

1S/O2/c1-2

InChI key

MYMOFIZGZYHOMD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Supplied in a carbon steel lecture bottle with a CGA180M/CGA110F needle valve installed.

Compatible with the following:
  • Aldrich® lecture-bottle station systems
  • Aldrich® lecture-bottle gas regulators

Other Notes

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

hose barb

Product No.
Description
Pricing

purge valve

Product No.
Description
Pricing

pictograms

Flame over circleGas cylinder

signalword

Danger

hcodes

Hazard Classifications

Ox. Gas 1 - Press. Gas Compr. Gas

Storage Class

2A - Gases

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ana Luísa Pinho et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(18), 6156-6163 (2014-05-03)
Musicians have been used extensively to study neural correlates of long-term practice, but no studies have investigated the specific effects of training musical creativity. Here, we used human functional MRI to measure brain activity during improvisation in a sample of
D K Heyland et al.
Critical care medicine, 24(3), 517-524 (1996-03-01)
To systemically review the effect of interventions designed to achieve supraphysiologic values of cardiac index, oxygen delivery (DO2), and oxygen consumption (VO2) in critically ill patients. Computerized bibliographic search of published research, citation review of relevant articles, and contact with
Anthony M-H Ho et al.
Chest, 123(3), 882-890 (2003-03-12)
To evaluate, by systematic review, the efficacy of heliox on respiratory mechanics and outcomes in patients with acute asthma. The search strategy included searching electronic databases (MEDLINE, EMBASE, and The Cochrane Library) and the references of relevant articles. Study quality
D R Spahn et al.
Anesthesia and analgesia, 78(5), 1000-1021 (1994-05-01)
Acute isovolemic hemodilution is used increasingly to avoid the potentially serious side effects of homologous blood transfusions. Cardiovascular physiology during hemodilution is characterized by a marked increase in cardiac output and organ blood flow to compensate for the decrease in
L Van Heerebeek et al.
Journal of clinical pathology, 55(8), 561-568 (2002-07-31)
Reactive oxygen species play an important role in a variety of (patho)physiological vascular processes. Recent publications have produced evidence of a role for putative non-phagocyte NADP oxidase(s) in the vascular production of reactive oxygen species. In the present review, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service