Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

29400

Sigma-Aldrich

D-α,β-Cyclohexylideneglycerol

≥98.0% (sum of enantiomers, GC)

Synonym(s):

(S)-(+)-1,4-Dioxaspiro[4.5]decane-2-methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16O3
CAS Number:
Molecular Weight:
172.22
Beilstein/REAXYS Number:
108000
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

≥98.0% (sum of enantiomers, GC)

form

liquid

optical activity

[α]20/D +5.9±0.5°, c = 5% in methanol

refractive index

n20/D 1.477 (lit.)
n20/D 1.480

bp

118-120 °C/5 mmHg (lit.)

density

1.096 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

OC[C@H]1COC2(CCCCC2)O1

InChI

1S/C9H16O3/c10-6-8-7-11-9(12-8)4-2-1-3-5-9/h8,10H,1-7H2/t8-/m0/s1

InChI key

XUGYZVQSZWPABZ-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

D-α,β-Cyclohexylideneglycerol can be used:
  • As an intermediate in the synthesis of antiarrhythmic oral drugs: (+)- and (−)-mexiletine.[1]
  • As a precursor in the preparation of glycoglycerolipids.[2]
  • As an intermediate in the synthesis of (S)-hydroxymethyl-1,4-benzodioxane, a key intermediate for the preparation of α-adrenergic receptor antagonists, and antidepressant drug doxazosin.[3]

Other Notes

Chiral building block; used e.g. in the synthesis of glycerides and carbohydrates[4]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A facile approach to chiral 1, 4-benzodioxane toward the syntheses of doxazosin, prosympal, piperoxan, and dibozane
Rouf A, et al.
Tetrahedron Letters, 54(48), 6420-6422 (2013)
R.R. Schmidt et al.
J. Carbohydr. Res., 3, 67-67 (1984)
Synthesis and thermotropic phase behavior of four glycoglycerolipids
Hogendorf WFJ, et al.
Molecules (Basel), 18(11), 13546-13573 (2013)
Enantiodivergent syntheses of (+)-and (−)-1-(2, 6-dimethylphenoxy) propan-2-ol: A way to access (+)-and (−)-mexiletine from D-(+)-mannitol
Manna A, et al.
Carbohydrate Research, 487, 107892-107892 (2020)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service