Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

293547

Sigma-Aldrich

exo-2-Chloronorbornane

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11Cl
CAS Number:
Molecular Weight:
130.62
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

refractive index

n20/D 1.4854 (lit.)

bp

66-66.5 °C/30 mmHg (lit.)

density

1.06 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

ClC1C[C@@H]2CC[C@H]1C2

InChI

1S/C7H11Cl/c8-7-4-5-1-2-6(7)3-5/h5-7H,1-4H2/t5-,6+,7?/m1/s1

InChI key

PJWBUKHIZPKRJF-JEAXJGTLSA-N

General description

exo-2-Chloronorbornane undergoes amination with trichloramine-aluminum chloride to yield 2-azabicyclo[3.2.1]octane, small amounts of the 3-isomer and exo-2-aminonorbornane[1].

Application

exo-2-Chloronorbornane has been employed as reagent during Friedel-Crafts alkylation of benzene and substituted benzenes with tin(IV) chloride or aluminium trichloride[2].

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

111.2 °F - closed cup

flash_point_c

44 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aromatic substitution. 64. Friedel-Crafts Alkylation of Aromatics with exo-2-Chloro-and 7-Chloronorbornane.
Olah GA, et al.
The Journal of Organic Chemistry, 59(9), 2590-2593 (1994)
Amination of< i> exo</i>-2-chloronorbornane and norbornane with trichloramine-aluminum chloride.
Kovacic P, et al.
Tetrahedron, 26(2), 529-538 (1970)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service