Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

293466

Sigma-Aldrich

Ethyl β-oxo-3-furanpropionate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O4
CAS Number:
Molecular Weight:
182.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

refractive index

n20/D 1.489 (lit.)

bp

113-116 °C/4 mmHg (lit.)

density

1.153 g/mL at 25 °C (lit.)

functional group

ester
ketone

SMILES string

CCOC(=O)CC(=O)c1ccoc1

InChI

1S/C9H10O4/c1-2-13-9(11)5-8(10)7-3-4-12-6-7/h3-4,6H,2,5H2,1H3

InChI key

OVPIQFGJQAQWAJ-UHFFFAOYSA-N

Application

Ethyl β-oxo-3-furanpropionate has been used in the synthesis of deuterated 4-ipomeanol[1].

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Teresa M Alvarez-Diez et al.
Drug metabolism and disposition: the biological fate of chemicals, 32(12), 1345-1350 (2004-08-26)
Earlier studies postulated that bioactivation of 4-ipomeanol by cytochrome P450 enzymes may occur through oxidation of its furan ring, following a mechanism similar to the bioactivation of other furan-containing compounds. This would lead to the formation of furan epoxides and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service