Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

291420

Sigma-Aldrich

2-Methoxypyrazine

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2O
CAS Number:
Molecular Weight:
110.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

refractive index

n20/D 1.509 (lit.)

bp

60-61 °C/29 mmHg (lit.)

density

1.14 g/mL at 25 °C (lit.)

SMILES string

COc1cnccn1

InChI

1S/C5H6N2O/c1-8-5-4-6-2-3-7-5/h2-4H,1H3

InChI key

WKSXRWSOSLGSTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

114.8 °F - closed cup

flash_point_c

46 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influence of pH and ageing on beer organoleptic properties. A sensory analysis based on AEDA data.
Guyot-Declerck C, et al.
Food Quality and Preference, 16(2), 157-162 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service