Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

291404

Sigma-Aldrich

Ethyl 3-phenylglycidate

technical grade, 92%, mixture of cis and trans

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12O3
CAS Number:
Molecular Weight:
192.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

assay

92%

refractive index

n20/D 1.518 (lit.)

bp

96 °C/0.5 mmHg (lit.)

density

1.102 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(=O)C1OC1c2ccccc2

InChI

1S/C11H12O3/c1-2-13-11(12)10-9(14-10)8-6-4-3-5-7-8/h3-7,9-10H,2H2,1H3

InChI key

GOMAKLPNAAZVCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Enantioselective epoxide hydrolase catalyzed resolution of ethyl 3-phenylglycidate by whole cells of Pseudomonas sp. has been investigated[1].

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Congfa Li et al.
Biotechnology letters, 25(24), 2113-2116 (2004-02-19)
A Pseudomonas sp. was isolated with enantioselective epoxide hydrolase activity to ethyl 3-phenylglycidate. Cells grown on sucrose and suspended in 10% (v/v) dimethyl formamide as co-solvent produced (2R,3S) ethyl 3-phenylglycidate with 95% ee and 26% yield in 12 h from
C Tilch et al.
Mutation research, 138(1), 1-8 (1984-10-01)
EPG and an in vitro digest of EPG by pepsin and pancreatin simulating mammalian digestion have been examined for genotoxicity in 4 mutagenicity tests employing different genetic endpoints. In the Salmonella reverse mutation assay, EPG showed only slight mutagenic activity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service