290769
2-Furaldehyde dimethylhydrazone
97%
Synonym(s):
Furfural dimethylhydrazone
About This Item
Recommended Products
assay
97%
refractive index
n20/D 1.578 (lit.)
bp
98 °C/9.5 mmHg (lit.)
density
1.042 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
CN(C)\N=C\c1ccco1
InChI
1S/C7H10N2O/c1-9(2)8-6-7-4-3-5-10-7/h3-6H,1-2H3/b8-6+
InChI key
DURWBNUASAZMSN-SOFGYWHQSA-N
General description
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
195.8 °F - closed cup
flash_point_c
91 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service