Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

290750

Sigma-Aldrich

1-(8-Quinolinesulfonyl)-3-nitro-1H-1,2,4-triazole

98%

Synonym(s):

QSNT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H7N5O4S
CAS Number:
Molecular Weight:
305.27
Beilstein/REAXYS Number:
4547109
MDL number:
UNSPSC Code:
41100000
PubChem Substance ID:
Pricing and availability is not currently available.

assay

98%

mp

215-217 °C (dec.)

SMILES string

[O-][N+](=O)c1ncn(n1)S(=O)(=O)c2cccc3cccnc23

InChI

1S/C11H7N5O4S/c17-16(18)11-13-7-15(14-11)21(19,20)9-5-1-3-8-4-2-6-12-10(8)9/h1-7H

InChI key

VFMMVUDTWOVVTC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service