Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28961

Sigma-Aldrich

1,3-Bis(cyanomethyl)imidazolium bis(trifluoromethylsulfonyl)imide

≥94% (HPLC/T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7F6N5O4S2
CAS Number:
Molecular Weight:
427.30
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥94% (HPLC/T)

form

solid

impurities

≤1.0% water

mp

81-85 °C

SMILES string

N#CCn1cc[n+](CC#N)c1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

InChI

1S/C7H7N4.C2F6NO4S2/c8-1-3-10-5-6-11(7-10)4-2-9;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h5-7H,3-4H2;/q+1;-1

InChI key

ORJULSCFWHFTFU-UHFFFAOYSA-N

Application

Nitrile-functionalized ionic liquid showing superior characteristics in Suzuki and Stille coupling reactions[1].

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongbin Zhao et al.
Journal of the American Chemical Society, 126(48), 15876-15882 (2004-12-02)
A series of relatively low-cost ionic liquids, based on the N-butyronitrile pyridinium cation [C(3)CNpy](+), designed to improve catalyst retention, have been prepared and evaluated in Suzuki and Stille coupling reactions. Depending on the nature of the anion, these salts react

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service