Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

288950

Sigma-Aldrich

Methyl 5-(benzyloxycarbonyl)-2,4-dimethyl-3-pyrrolepropionate

97%

Synonym(s):

Methyl 2,4-dimethyl-5-[(phenylmethoxy)carbonyl]-1H-pyrrole-3-propanoate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$356.00

$356.00


Available to ship onMay 05, 2025Details


Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
5 G
$356.00

About This Item

Empirical Formula (Hill Notation):
C18H21NO4
CAS Number:
Molecular Weight:
315.36
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$356.00


Available to ship onMay 05, 2025Details


Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

assay

97%

form

powder

mp

98-99 °C (lit.)

functional group

ester
phenyl

SMILES string

COC(=O)CCc1c(C)[nH]c(C(=O)OCc2ccccc2)c1C

InChI

1S/C18H21NO4/c1-12-15(9-10-16(20)22-3)13(2)19-17(12)18(21)23-11-14-7-5-4-6-8-14/h4-8,19H,9-11H2,1-3H3

Inchi Key

KCXNIAMRWFWMOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl 5-(benzyloxycarbonyl)-2,4-dimethyl-3-pyrrolepropionate may be used in chemical synthesis studies.

Pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service