Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

287334

Sigma-Aldrich

Triamcinolone

Synonym(s):

9α-Fluoro-11β,16α,17,21-tetrahydroxy-1,4-pregnadiene-3,20-dione, 9α-Fluoro-11β,16α,17,21-tetrahydroxypregna-1,4-diene-3,20-dione, Fluoxyprednisolone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27FO6
CAS Number:
Molecular Weight:
394.43
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

optical activity

[α]25/D +69°, c = 2 in DMF

mp

262-263 °C (lit.)

SMILES string

[H][C@@]12C[C@@H](O)[C@](O)(C(=O)CO)[C@@]1(C)C[C@H](O)[C@@]3(F)[C@@]2([H])CCC4=CC(=O)C=C[C@]34C

InChI

1S/C21H27FO6/c1-18-6-5-12(24)7-11(18)3-4-13-14-8-15(25)21(28,17(27)10-23)19(14,2)9-16(26)20(13,18)22/h5-7,13-16,23,25-26,28H,3-4,8-10H2,1-2H3/t13-,14-,15+,16-,18-,19-,20-,21-/m0/s1

InChI key

GFNANZIMVAIWHM-OBYCQNJPSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Triamcinolone is a synthetic glucocorticoid agonist; induces gene expression and apoptosis; inhibits prostaglandin synthesis; impairs tumor necrosis factor (TNF)-α-induced degradation of κB-α; potentiates the differentiation-inducing effects of bone morphogenetic proteins (BMP-2, -4, -6).

replaced by

Product No.
Description
Pricing

pictograms

Health hazard

signalword

Warning

hcodes

pcodes

Hazard Classifications

Carc. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wai-Leung Langston Suen et al.
Journal of controlled release : official journal of the Controlled Release Society, 167(1), 21-28 (2013-01-15)
We are proposing folate-decorated polymeric nanoparticles as carriers of poorly soluble drug molecules for intracellular and prolonged delivery to retinal pigment epithelium (RPE) cells. RPE is a monolayer of epithelial cells that forms the outer blood-retinal barrier in the posterior
Thøger Persson Krogh et al.
The American journal of sports medicine, 41(3), 625-635 (2013-01-19)
Lateral epicondylitis (LE) is a common musculoskeletal disorder for which an effective treatment strategy remains unknown. To examine whether a single injection of platelet-rich plasma (PRP) is more effective than placebo (saline) or glucocorticoid in reducing pain in adults with
Andrew Y H Chin et al.
The Journal of hand surgery, 38(2), 316-321 (2013-01-09)
We report a series of 18 patients with chronic sesamoiditis of the metacarpophalangeal joint of the thumb treated in the period from 1997 to 2002. We describe a diagnostic clinical test (sesamoid provocation test [SPT]) and present the long-term results
Mariana Figueiroa Careta et al.
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.], 39(5), 734-738 (2013-02-06)
Earlobe keloids are benign, fibrous proliferations that occur in predisposed persons at sites of cutaneous injury. No single best therapeutic modality is indicated. To describe a 1-year follow-up of 12 patients with earlobe keloids treated by shaving followed by cryosurgery
Ali Shakir et al.
American journal of physical medicine & rehabilitation, 92(9), 768-775 (2013-02-02)
This study aimed to compare the effectiveness of triamcinolone vs. dexamethasone used in transforaminal epidural steroid injections for the treatment of cervical radiculopathy. This is a retrospective cohort study of patients with cervical radiculopathy who underwent cervical transforaminal epidural steroid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service