Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

287164

Sigma-Aldrich

Spermine tetrahydrochloride

98%

Synonym(s):

N,N′-Bis(3-Aminopropyl)-1,4-butanediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)3NH(CH2)4NH(CH2)3NH2 · 4HCl
CAS Number:
Molecular Weight:
348.18
Beilstein/REAXYS Number:
3911771
UNSPSC Code:
12352103
PubChem Substance ID:

assay

98%

InChI

1S/C10H26N4.ClH/c11-5-3-9-13-7-1-2-8-14-10-4-6-12;/h13-14H,1-12H2;1H

InChI key

KBDDIZRDKLGWGW-UHFFFAOYSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong Kee Choi et al.
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology, 19(2), 77-84 (2008-10-11)
Levels of ionotropic glutamate (Glu) N-methyl-d-aspartic acid (NMDA), 2-amino-3-(3-hydroxy-5-methyl-isoxazol-4-yl)propionic acid (AMPA), and kainic acid (KA) receptors in forebrain regions of juvenile rats (age 42 days) were quantified after 3 weeks of treatment with three different doses of risperidone (0.3, 1.0
Enzo Agostinelli et al.
International journal of oncology, 45(3), 1109-1122 (2014-06-28)
It has been confirmed that multidrug resistant (MDR) melanoma cells (M14 ADR2) are more sensitive than their wild-type counterparts (M14 WT) to H2O2 and aldehydes, the products of bovine serum amine oxidase (BSAO)-catalyzed oxidation of spermine. The metabolites formed by
Desiree Bailey et al.
Pharmacology, biochemistry, and behavior, 133, 57-64 (2015-04-01)
The aim of this study was to examine the acute effect of a range of novel hydroxycinnamic acid derivatives of spermine on the development of spermine-induced CNS excitation and convulsions in female Laca mice, and to assess the chronic adverse
Xinna Wang et al.
Biomedical chromatography : BMC, 32(6), e4211-e4211 (2018-02-16)
We describe a simple, rapid, selective and sensitive HPLC method coupled with fluorescence detection for simultaneous determination of 10 kinds of biogenic amines (BAs: tryptamine, 2-phenethylamine, putrescine, cadaverine, histamine, 5-hydroxytryptamine, tyramine, spermidine, dopamine and spermine). BAs and IS were derivated
Alexis Goichon et al.
The American journal of clinical nutrition, 102(2), 359-367 (2015-06-26)
Amino acids are well known to be key effectors of gut protein turnover. We recently reported that enteral delivery of proteins markedly stimulated global duodenal protein synthesis in carbohydrate-fed healthy humans, but specifically affected proteins remain unknown. We aimed to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service