Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28625

Sigma-Aldrich

Cyanuric fluoride

Synonym(s):

2,4,6-Trifluoro-1,3,5-triazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3F3N3
CAS Number:
Molecular Weight:
135.05
Beilstein/REAXYS Number:
124237
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

SMILES string

Fc1nc(F)nc(F)n1

InChI

1S/C3F3N3/c4-1-7-2(5)9-3(6)8-1

InChI key

VMKJWLXVLHBJNK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

can precipitate on storage

Other Notes

Modifies tyrosyl residues in proteins[1][2]; Reagent used for the preparation of carboxylic fluorides[3][4]

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Eye Dam. 1 - Skin Corr. 1A

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

[43] cyanuration.
M J Gorbunoff
Methods in enzymology, 25, 506-514 (1972-01-01)
Tyrosyl residues of two molecular forms of human urinary urokinase: their ionization mode characterized by spectrophotometric and CD titrations and reactivity with cyanuric fluoride in relation to enzymic activity.
N Miwa et al.
Biochemical and biophysical research communications, 108(3), 1136-1143 (1982-10-15)
G.A. Olah et al.
Synthesis, 487-487 (1973)
M.J.S. Dewar et al.
The Journal of Organic Chemistry, 40, 1521-1521 (1975)
Young Shik Chi et al.
Langmuir : the ACS journal of surfaces and colloids, 23(3), 1209-1214 (2007-01-24)
This report describes the reactivity of acid fluoride (AF)-terminated self-assembled monolayers (SAMs) on gold toward amine and alcohol compounds and the potentiality of AF as a reactive intermediate for surface functionalizations. The AF group was generated in situ on a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service