Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

285803

Sigma-Aldrich

Estriol

98%

Synonym(s):

1,3,5(10)-Estratriene-3,16α,17β-triol, 16α-Hydroxyestradiol, 3,16α,17β-Trihydroxy-1,3,5(10)-estratriene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24O3
CAS Number:
Molecular Weight:
288.38
Beilstein/REAXYS Number:
2508172
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:

assay

98%

form

solid

optical activity

[α]20/D +62°, c = 1 in 95% ethanol

mp

280-282 °C (lit.)

SMILES string

C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1C[C@@H](O)[C@@H]2O

InChI

1S/C18H24O3/c1-18-7-6-13-12-5-3-11(19)8-10(12)2-4-14(13)15(18)9-16(20)17(18)21/h3,5,8,13-17,19-21H,2,4,6-7,9H2,1H3/t13-,14-,15+,16-,17+,18+/m1/s1

InChI key

PROQIPRRNZUXQM-ZXXIGWHRSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Health hazard

signalword

Danger

Hazard Classifications

Carc. 2 - Lact. - Repr. 1A

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J J Evans et al.
Clinical chemistry, 30(1), 120-121 (1984-01-01)
Saliva would have advantages over plasma or urine for monitoring estriol during pregnancy. Specimen collection, after stimulation of flow by citric acid, is non-invasive and simple. We measured concentrations of unconjugated estriol in saliva and compared them with those in
E Diczfalusy
Journal of steroid biochemistry, 20(4B), 945-953 (1984-04-01)
The early history of estriol is reviewed with special emphasis on its isolation, identification, quantitation in tissues and body fluids and its unique biogenesis in the human feto-placental unit. The relationship to epimeric estriols and acidic estrogens and the pharmacokinetic
Nina Sneitz et al.
Drug metabolism and disposition: the biological fate of chemicals, 41(3), 582-591 (2013-01-05)
The glucuronidation of estriol, 16-epiestriol, and 17-epiestriol by the human UDP-glucuronosyltransferases (UGTs) of subfamilies 1A, 2A, and 2B was examined. UGT1A10 is highly active in the conjugation of the 3-OH in all these estriols, whereas UGT2B7 is the most active
Hanna Hamid et al.
Water research, 46(18), 5813-5833 (2012-09-04)
Estrogenic hormones (estrone (E1), 17β-estradiol (E2), estriol (E3), 17α-ethinylestradiol (EE2)) are the major contributor to the total estrogenicity in waterways. Presence of these compounds in biosolids is also causing concern in terms of their use as soil amendment. In comparison
Marcelo O Dietrich et al.
Trends in neurosciences, 36(2), 65-73 (2013-01-16)
The past 20 years witnessed an enormous leap in understanding of the central regulation of whole-body energy metabolism. Genetic tools have enabled identification of the region-specific expression of peripheral metabolic hormone receptors and have identified neuronal circuits that mediate the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service