Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

285153

Sigma-Aldrich

Aniline sulfate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5NH2)2 · H2SO4
CAS Number:
Molecular Weight:
284.33
Beilstein/REAXYS Number:
3729533
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

98%

solubility

water: soluble 5%, clear, colorless

SMILES string

OS(O)(=O)=O.Nc1ccccc1.Nc2ccccc2

InChI

1S/2C6H7N.H2O4S/c2*7-6-4-2-1-3-5-6;1-5(2,3)4/h2*1-5H,7H2;(H2,1,2,3,4)

InChI key

FUKMEFZGEMVGLD-UHFFFAOYSA-N

Application

Aniline sulfate was used as an internal standard in the stereoselective disposition of methamphetamine (MAP, a widely abused drug) study[1].

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Skin Sens. 1 - STOT RE 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Hutchaleelaha et al.
Journal of chromatography. B, Biomedical applications, 658(1), 103-112 (1994-08-05)
In order to study the stereoselective disposition of methamphetamine (MAP), a widely abused drug, we have developed a sensitive HPLC assay to separate and quantitate the enantiomers of MAP and amphetamine (AP) in rat serum. Serum samples to which was
Vani Nagaraja et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 173, 407-417 (2016-10-05)
Spectrophotometric method with three systems were developed here for the determination of gold(III) using o-dianisidine, aniline sulphate and catechol. Gold(III),in the system 1 it oxidizes o-dianisidine, in the system 2 it oxidizes catechol followed by its coupling with o-dianisidine, in
Conghui Tang et al.
Journal of the American Chemical Society, 134(46), 18924-18927 (2012-11-08)
A novel and efficient copper-catalyzed azidation reaction of anilines via C-H activation has been developed. This method, in which the primary amine acts as a directing group by coordinating to the metal center, provides ortho azidation products regioselectively under mild
Huihui Yang et al.
Electrophoresis, 34(4), 510-517 (2012-11-23)
A novel (3-sulfopropyl methacrylate potassium)-silica hybrid monolithic column for CEC has been prepared by a simple one-pot approach based on efficient thiol-ene click chemistry. In this process, the polycondensation of hydrolyzed alkoxysilanes and in situ click reaction of vinyl groups
Dongdong Liang et al.
Chemical communications (Cambridge, England), 49(2), 173-175 (2012-11-22)
An efficient synthesis of free (NH)-phenanthridinones through Pd-catalyzed C(sp(2))-H aminocarbonylation of unprotected o-arylanilines under an atmospheric pressure of CO has been developed. Some ortho heteroarene substituted anilines as well as N-alkyl protected o-arylanilines are also suitable substrates for this C-H

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service