Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

284319

Sigma-Aldrich

1,8-Dinitropyrene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H8N2O4
CAS Number:
Molecular Weight:
292.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

98%

solubility

DMSO: soluble 2 mg/mL, clear, faintly yellow to yellow (faint green-yellow to very dark)

SMILES string

[O-][N+](=O)c1ccc2ccc3ccc([N+]([O-])=O)c4ccc1c2c34

InChI

1S/C16H8N2O4/c19-17(20)13-7-3-9-1-2-10-4-8-14(18(21)22)12-6-5-11(13)15(9)16(10)12/h1-8H

InChI key

BLYXNIHKOMELAP-UHFFFAOYSA-N

General description

The photodecomposition of environmental contaminant, 1,8-dinitropyrene via exposure to light (≥ 310nm) either in DMSO, or following coating onto silica has been investigated[1]. 1,8-Dinitropyrene is a potent mutagenic component of diesel exhaust and air pollutant[2].

pictograms

Health hazard

signalword

Warning

hcodes

pcodes

Hazard Classifications

Carc. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P J Harris et al.
Mutation research, 412(3), 323-331 (1998-05-26)
The abilities of brans from the cereals barley, oats, maize, rice, and wheat to adsorb in vitro the hydrophobic, environmental mutagen 1,8-dinitropyrene (DNP) were investigated using a mutagenicity assay. These brans were obtained from known cultivars using defined milling conditions
T Nohmi et al.
Molecular & general genetics : MGG, 247(1), 7-16 (1995-04-10)
Expression of the umuDC operon is required for UV and most chemical mutagenesis in Escherichia coli. The closely related species Salmonella typhimurium has two sets of umuDC-like operons, umuDCST on the chromosome and samAB on a 60-MDa cryptic plasmid. The
M A Belisario et al.
Bollettino della Societa italiana di biologia sperimentale, 71(1-2), 27-34 (1995-01-01)
Nitroarenes are environmental contaminants produced during incomplete combustion processes. It has been reported that nitroreduction, the most important pathway of nitroarene toxification, occurs mainly in the liver and intestine. In the present study we have shown that red cells also
L R Ferguson et al.
Chemico-biological interactions, 95(3), 245-255 (1995-04-14)
Epidemiology and animal experiments indicate that dietary fibres protect against the development of colorectal cancer. However, insoluble dietary fibres appear to be more effective than soluble dietary fibres and one mechanism by which they may protect is by adsorbing dietary
Nitroarenes (selected) 1,8-dinitropyrene.
Report on carcinogens : carcinogen profiles, 11, III187-III188 (2004-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service