Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

283150

Sigma-Aldrich

2,4,6-Trimethylpyridinium p-toluenesulfonate

98%

Synonym(s):

p-Toluenesulfonic acid sym-Collidine salt, sym-Collidinium p-toluenesulfonate, CTPS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H19NO3S
CAS Number:
Molecular Weight:
293.38
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

solid

mp

128-130 °C (lit.)

functional group

tosylate

SMILES string

Cc1cc(C)nc(C)c1.Cc2ccc(cc2)S(O)(=O)=O

InChI

1S/C8H11N.C7H8O3S/c1-6-4-7(2)9-8(3)5-6;1-6-2-4-7(5-3-6)11(8,9)10/h4-5H,1-3H3;2-5H,1H3,(H,8,9,10)

InChI key

VEXWNPGPVMYVDU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,4,6-Trimethylpyridinium p-toluenesulfonate may be used in chemical synthesis.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bo Zhang et al.
Journal of genetics and genomics = Yi chuan xue bao, 47(3), 131-143 (2020-04-23)
Compartmentation of enzymes via filamentation has arisen as a mechanism for the regulation of metabolism. In 2010, three groups independently reported that CTP synthase (CTPS) can assemble into a filamentous structure termed the cytoophidium. In searching for CTPS-interacting proteins, here
Eric M Lynch et al.
Nature structural & molecular biology, 27(1), 42-48 (2019-12-25)
Many enzymes assemble into defined oligomers, providing a mechanism for cooperatively regulating activity. Recent studies have described a mode of regulation in which enzyme activity is modulated by polymerization into large-scale filaments. Here we describe an ultrasensitive form of polymerization-based
Gregory D McCluskey et al.
Journal of molecular biology, 430(8), 1201-1217 (2018-03-05)
While enzyme activity is often regulated by a combination of substrate/effector availability and quaternary structure, many cytosolic enzymes may be further regulated through oligomerization into filaments. Cytidine-5'-triphosphate (CTP) synthase (CTPS) forms such filaments-a process that is promoted by the product
Zhe Sun et al.
Journal of genetics and genomics = Yi chuan xue bao, 46(2), 65-74 (2019-03-13)
CTP synthase (CTPS), the rate-limiting enzyme in de novo CTP biosynthesis, has been demonstrated to assemble into evolutionarily conserved filamentous structures, termed cytoophidia, in Drosophila, bacteria, yeast and mammalian cells. However, the regulation and function of the cytoophidium remain elusive.
Anne-Claire Boschat et al.
Journal of mass spectrometry : JMS, 54(11), 885-893 (2019-09-17)
Cytidine 5'-triphosphate synthetase (CTPS) is known to be a central enzyme in the de novo synthesis of CTP. We have recently demonstrated that a deficiency in CTPS1 is associated with an impaired capacity of activated lymphocytes to proliferate leading to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service