Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

278793

Sigma-Aldrich

1-Methylpiperazine dihydrochloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12N2 · 2HCl
CAS Number:
Molecular Weight:
173.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

98%

form

solid

mp

246-250 °C (lit.)

solubility

1 M NH4OH: soluble 25 mg/mL, clear to hazy, colorless to light yellow

SMILES string

Cl[H].Cl[H].CN1CCNCC1

InChI

1S/C5H12N2.2ClH/c1-7-4-2-6-3-5-7;;/h6H,2-5H2,1H3;2*1H

InChI key

AILFRWRYZZVJTL-UHFFFAOYSA-N

Application

1-Methylpiperazine dihydrochloride (Methylpiperazine dihydrochloride) has been used in the preparation of 1-methylpiperazine-1,4-diium tetrachloridozincate hemihydrate[1].

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sondra Walha et al.
Acta crystallographica. Section E, Structure reports online, 67(Pt 11), m1605-m1605 (2012-01-06)
The crystal structure of the title compound, (C(5)H(14)N(2))[ZnCl(4)]·0.5H(2)O, is built up from discrete 1-methyl-piperazine-diium cations with chair conformation, tetrahedral tetrachloridozincate anions and uncoordinated solvent water mol-ecules linked together by three types of inter-molecular hydrogen bonds, viz. N-H⋯Cl, N-H⋯O and O-H⋯Cl.
H Welinder et al.
International archives of allergy and applied immunology, 79(3), 259-262 (1986-01-01)
The presence of specific IgE antibodies against conjugates between human serum albumin and piperazine, a compound previously known to cause occupational asthma, and N-methyl-piperazine, respectively, was demonstrated by RAST, RAST inhibition and skin prick tests in 2 asthmatic subjects occupationally
J M Entrena et al.
Scientific reports, 6, 37835-37835 (2016-11-26)
Sigma-1 receptor antagonists promote antinociception in several models of pain, but the effects of sigma-1 agonists on nociception (particularly when the nociceptive system is primed) are not so well characterized; therefore we evaluated the effects of sigma-1 agonists on pain
E Gavathiotis et al.
Nucleic acids research, 28(3), 728-735 (2000-01-19)
The solution structure of the dodecamer duplex d(CTTTTGCAAAAG)(2)and its 2:1 complex with the bis -benzimidazole Hoechst 33258 has been investigated by NMR and NOE-restrained molecular dynamics (rMD) simulations. Drug molecules are bound in each of the two A-tracts with the
Olesya A Troshina et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 12(21), 5569-5577 (2006-06-07)
An oxidative radical photoaddition of mono N-substituted piperazines to [60]fullerene was systematically investigated. Reactions of C60 with piperazines bearing bulky electron-withdrawing groups (2-pyridyl, 2-pyrimidinyl) were found to be the most selective and yielded C60(amine)4O as major products along with small

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service