Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

277673

Sigma-Aldrich

Benzyl 4-nitrophenyl carbonate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4OCO2CH2C6H5
CAS Number:
Molecular Weight:
273.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

99%

mp

78-80 °C (lit.)

functional group

carbonate
nitro
phenyl

SMILES string

[O-][N+](=O)c1ccc(OC(=O)OCc2ccccc2)cc1

InChI

1S/C14H11NO5/c16-14(19-10-11-4-2-1-3-5-11)20-13-8-6-12(7-9-13)15(17)18/h1-9H,10H2

InChI key

QIXRWIVDBZJDGD-UHFFFAOYSA-N

Application

Benzyl 4-nitrophenyl carbonate may be used in chemical synthesis.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service