Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

277320

Sigma-Aldrich

1,2-Dibromocyclopentene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6Br2
CAS Number:
Molecular Weight:
225.91
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

refractive index

n20/D 1.5558 (lit.)

bp

78 °C/5 mmHg (lit.)

density

1.895 g/mL at 25 °C (lit.)

functional group

bromo

storage temp.

2-8°C

SMILES string

BrC1=C(Br)CCC1

InChI

1S/C5H6Br2/c6-4-2-1-3-5(4)7/h1-3H2

InChI key

PNWFXPGGROADNS-UHFFFAOYSA-N

General description

1,2-Dibromocyclopentene undergoes single Br/Mg exchange reaction with iPrMgCl LiCl to yield the corresponding β-bromocyclopentenylmagnesium reagent[1].

Application

1,2-Dibromocyclopentene has been used in the synthesis of cyclopentene analogs[2].

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christina Despotopoulou et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 14(8), 2499-2506 (2008-01-31)
A single Br/Mg exchange of 1,2-dibromocyclopentene with iPrMgCl LiCl provides the corresponding beta-bromocyclopentenylmagnesium reagent, which can then be reacted with various electrophiles (yields: 65-82 %). In the presence of a secondary alkylmagnesium halide and Li2CuCl4 (2 mol %), these 2-bromoalkenylmagnesium
Gerard M P Giblin et al.
Bioorganic & medicinal chemistry letters, 17(2), 385-389 (2006-11-07)
The discovery of a series of selective EP1 receptor antagonists based on a 1,2-diarylcyclopentene template is described. After defining the structural requirements for EP1 potency and selectivity, heterocyclic rings were incorporated to reduce logD and improve in vitro pharmacokinetic properties.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service