Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

277061

Sigma-Aldrich

(S)-(−)-2-(Methoxymethyl)-1-pyrrolidinecarboxaldehyde

98%

Synonym(s):

(S)-(−)-1-Formyl-2-(methoxymethyl)pyrrolidine, (S)-(−)-2-(Methoxymethyl)pyrrolidine-1-carboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13NO2
CAS Number:
Molecular Weight:
143.18
Beilstein/REAXYS Number:
4377661
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

liquid

optical activity

[α]20/D −34°, c = 2 in toluene

refractive index

n20/D 1.4755 (lit.)

bp

67 °C/0.25 mmHg (lit.)

density

1.057 g/mL at 25 °C (lit.)

functional group

ether

storage temp.

2-8°C

SMILES string

COC[C@@H]1CCCN1C=O

InChI

1S/C7H13NO2/c1-10-5-7-3-2-4-8(7)6-9/h6-7H,2-5H2,1H3/t7-/m0/s1

InChI key

JNIOQRWRORXADR-ZETCQYMHSA-N

Application

Chiral auxiliary reagent for asymmetric C-C coupling.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service