Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

276650

Sigma-Aldrich

1-Chloro-1-cyclopentene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7Cl
CAS Number:
Molecular Weight:
102.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

refractive index

n20/D 1.4651 (lit.)

bp

113-115 °C (lit.)

density

1.035 g/mL at 25 °C (lit.)

functional group

chloro

storage temp.

2-8°C

SMILES string

ClC1=CCCC1

InChI

1S/C5H7Cl/c6-5-3-1-2-4-5/h3H,1-2,4H2

InChI key

UJUIJZWQFDQKHO-UHFFFAOYSA-N

General description

1-Chloro-1-cyclopentene undergoes cross-coupling reaction with arylboronic acids, arylzinc reagents and thiols catalyzed by air-stable palladium complexes to yield the corresponding styrene derivatives, biaryls, and thioethers[1]. Palladium-catalyzed aminocarbonylations of 1-chloro-1-cyclopentene and benzyl amine has been studied[2].

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

48.2 °F - closed cup

flash_point_c

9 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aminocarbonylations of alkenyl phosphates, chlorides, bromides, and triflates with Mo(CO)6 as a solid CO source.
Lagerlund O, et al.
Tetrahedron, 65(36), 7646-7652 (2009)
George Y Li
The Journal of organic chemistry, 67(11), 3643-3650 (2002-05-25)
Air-stable palladium complexes [(t-Bu)(2)P(OH)](2)PdCl(2), [(t-Bu)(2)P(OH)PdCl(2)](2), and [[(t-Bu)(2)PO...H...OP((t-Bu)(2)]PdCl](2) serve as efficient catalysts for a variety of cross-coupling reactions of vinyl and aryl chlorides with arylboronic acids, arylzinc reagents, and thiols to yield the corresponding styrene derivatives, biaryls, and thioethers. (31)P NMR

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service