Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

274011

Sigma-Aldrich

Copper(II) phthalocyanine-tetrasulfonic acid tetrasodium salt

Dye content ~50 %

Synonym(s):

Phthalocyanine-tetrasulfonic acid copper(II) complex tetrasodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H12CuN8O12S4Na4
CAS Number:
Molecular Weight:
984.25
Beilstein/REAXYS Number:
9609904
EC Number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

powder

composition

Dye content, ~50%

λmax

609 nm
692 nm (2nd)

SMILES string

[Na+].[Na+].[Na+].[Na+].[O-]S(=O)(=O)c1ccc2c3nc(nc4n5[Cu]n6c(nc7nc(nc5c8cc(ccc48)S([O-])(=O)=O)c9cc(ccc79)S([O-])(=O)=O)c%10ccc(cc%10c6n3)S([O-])(=O)=O)c2c1

InChI

1S/C32H16N8O12S4.Cu.4Na/c41-53(42,43)13-1-5-17-21(9-13)29-33-25(17)37-30-22-10-14(54(44,45)46)2-6-18(22)27(34-30)39-32-24-12-16(56(50,51)52)4-8-20(24)28(36-32)40-31-23-11-15(55(47,48)49)3-7-19(23)26(35-31)38-29;;;;;/h1-12H,(H4-2,33,34,35,36,37,38,39,40,41,42,43,44,45,46,47,48,49,50,51,52);;;;;/q-2;+2;4*+1/p-4

InChI key

HMPXSWYLSSRSQF-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J J Abramson et al.
Archives of biochemistry and biophysics, 263(2), 245-255 (1988-06-01)
The copper containing phthalocyanine dyes, alcian blue, copper phthalocyanine tetrasulfonic acid, and Luxol fast blue MBSN are found to induce rapid calcium efflux from actively loaded sarcoplasmic reticulum (SR) vesicles. Alcian blue (5 microM), with 1 mM free Mg2+ triggered
I K O'Neill et al.
Carcinogenesis, 11(4), 599-607 (1990-04-01)
Trapping by magnetic polyethyleneimine (PEI) microcapsules was utilized to investigate the influence in male rats of dose, human dietary composition and time-dependence on reactive metabolites of benzo[a]pyrene (B[a]P) in the gastrointestinal (GI) tract; also, PEI microcapsules modified with copper phthalocyanine
D Bickar et al.
Analytical biochemistry, 203(1), 109-115 (1992-05-15)
A method for protein staining using copper phthalocyanine 3,4',4'',4'''-tetrasulfonic acid tetrasodium salt is described. The procedure is applicable to protein blots and tissue prints, as well as to polyacrylamide and agarose gels. It is also simple, involving only application of
H Hayatsu
Journal of chromatography, 597(1-2), 37-56 (1992-04-24)
A method useful as a preconcentration technique for isolating mutagens and carcinogens is described. Cotton bearing covalently linked copper phthalocyanine trisulphonate as ligand (blue cotton) can adsorb selectively compounds having three or more fused rings. The adsorption takes place in
Hanming Ding et al.
Journal of colloid and interface science, 290(1), 166-171 (2005-06-21)
Tetrasulfonated copper phthalocyanine (CuTsPc)-capped TiO2 nanoparticle ultrathin films were fabricated by a layer-by-layer (LBL) self-assembly technique. Alternating bilayer structures were formed by consecutive adsorption of CuTsPc-capped TiO2 nanoparticles with poly(diallyldimethylammonium chloride). Optical and electrical measurements were carried out to characterize

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service