Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

271535

Sigma-Aldrich

Methyl all-cis-5,8,11,14,17-eicosapentaenoate

technical grade, ≥90%

Synonym(s):

cis-5,8,11,14,17-Eicosapentaenoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2CH=CH)5(CH2)3CO2CH3
CAS Number:
Molecular Weight:
316.48
Beilstein/REAXYS Number:
1914828
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

grade

technical grade

assay

≥90%

SMILES string

CC\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CCCC(=O)OC

InChI

1S/C21H32O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21(22)23-2/h4-5,7-8,10-11,13-14,16-17H,3,6,9,12,15,18-20H2,1-2H3/b5-4-,8-7-,11-10-,14-13-,17-16-

InChI key

QWDCYFDDFPWISL-JEBPEJKESA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service