Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

269867

Sigma-Aldrich

Nitromethane-d3

≥99 atom % D, contains 1 % (v/v) TMS

Synonym(s):

Trideuteronitromethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3NO2
CAS Number:
Molecular Weight:
64.06
Beilstein/REAXYS Number:
1751076
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:

isotopic purity

≥99 atom % D

assay

≥99.0%

form

liquid

contains

1 % (v/v) TMS

expl. lim.

7.1-63 % (lit.)

technique(s)

NMR: suitable

refractive index

n20/D 1.3795 (lit.)

bp

100 °C (lit.)

mp

-29 °C (lit.)

density

1.183 g/mL at 25 °C (lit.)

mass shift

M+3

SMILES string

[2H]C([2H])([2H])[N+]([O-])=O

InChI

1S/CH3NO2/c1-2(3)4/h1H3/i1D3

Inchi Key

LYGJENNIWJXYER-FIBGUPNXSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 3 - Repr. 2

Storage Class

4.1A - Other explosive hazardous materials

wgk_germany

WGK 2

flash_point_f

95.0 °F - closed cup

flash_point_c

35 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isotope effects on selected physicochemical properties of nitromethane and 1-pentanol.
Makowska A and Szydlowski J.
Nukleonika, 51, 81-85 (2006)
DFT molecular structure and initial assignment for the FTIR spectrum of ?7 band of nitromethane-D3.
Pal C and Kshirsagar RJ.
Indian J. Phys., 84(8), 929-937 (2010)
Pyrolysis of nitromethane-d3.
Crawforth CG and Waddington DJ.
The Journal of Physical Chemistry, 74(4), 2793-2796 (1970)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service