Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

269735

Sigma-Aldrich

Cyclohexane-d12

≥99.6 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D12
CAS Number:
Molecular Weight:
96.23
Beilstein/REAXYS Number:
1908842
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:

isotopic purity

≥99.6 atom % D

assay

≥99% (CP)

form

liquid

expl. lim.

0.34-6.3 % (lit.)

packaging

pkg of 10 × 1 mL

technique(s)

NMR: suitable

refractive index

n20/D 1.421 (lit.)

bp

80.7 °C (lit.)

mp

6.5 °C (lit.)

density

0.893 g/mL at 25 °C (lit.)

mass shift

M+12

SMILES string

[2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])[2H]

InChI

1S/C6H12/c1-2-4-6-5-3-1/h1-6H2/i1D2,2D2,3D2,4D2,5D2,6D2

InChI key

XDTMQSROBMDMFD-LBTWDOQPSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

-0.4 °F - closed cup

flash_point_c

-18 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A vibrational study of cyclohexane and some of its isotopic derivatives-I: Raman and infrared spectra and assignments of cyclohexane and cyclohexane-d12.
Wiberg KB and Shrake A.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 27(7), 1139-1151 (1971)
Dynamic deuterium NMR in liquid crystalline solvents: Ring inversion of cyclohexane-d12.
Poupko R and Luz Z.
J. Chem. Phys. , 75(4), 1675-1681 (1981)
Viscosities of benzene-d6 and cyclohexane-d12.
Dixon JA and Schiessler RW.
The Journal of Physical Chemistry, 58(5), 430-432 (1954)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service