Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

269034

Sigma-Aldrich

Methanesulfonamide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SO2NH2
CAS Number:
Molecular Weight:
95.12
Beilstein/REAXYS Number:
1740835
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

98%

mp

85-89 °C (lit.)

SMILES string

CS(N)(=O)=O

InChI

1S/CH5NO2S/c1-5(2,3)4/h1H3,(H2,2,3,4)

InChI key

HNQIVZYLYMDVSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sham M Sondhi et al.
Molecular diversity, 13(3), 357-366 (2009-03-10)
Condensation of 3,4-diaryl-2-imino-4-thiazoli-nes 1a-j with methanesulfonyl chloride gave methanesulfonamide derivatives 2a-j. Condensation of 2-cyanopyrazine, 4-cyanopyridine, and 2-cyanopyridine with 3,4-diaryl-2- imino-4-thiazolines 1h-m in the presence of sodium methoxide afforded amidine derivatives 3a-j. Compounds 2a-j and 3a-j were screened against various human
Yoshimichi Shoji et al.
Bioorganic & medicinal chemistry, 17(16), 6020-6031 (2009-07-18)
A novel series of indoline-based acyl-CoA: cholesterol acyltransferase (ACAT) inhibitors with a methanesulfonamide group at the 5-position were synthesized and their lipophilicity and biological activities were evaluated. Hepatic ACAT inhibitory and anti-foam cell formation activity increased dependent on lipophilicity of
Jonathan B Grimm et al.
The Journal of organic chemistry, 72(21), 8135-8138 (2007-09-21)
An efficient two-step strategy has been developed to access diversely functionalized benzylic sulfonamides. Execution of this strategy required the development of two reaction methods: the palladium-catalyzed cross-coupling of aryl halides with CH-acidic methanesulfonamides and a metathesis reaction between the resulting
Kei Ohara et al.
International journal of colorectal disease, 18(3), 260-266 (2003-04-04)
We recently reported the phenomenon of prostaglandin-dependent colonic adaptive cytoprotection (CAP) against acute colonic injury induced by acetic acid (AA) in the normal colon. This study investigated whether the CAP is preserved in the chronic inflamed colon. Normal rats and
Hamit Alyar et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 91, 39-47 (2012-02-22)
A conformational analysis of the methanesulfonamide-N,N'-1,2-ethanediylbis (msen) was performed by using vibrational and NMR spectroscopies as well as theoretical computations. The possible stable conformers of msen on its potential energy surface were investigated by semi-empirical PM5 method and appropriate structures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service