Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

266795

Sigma-Aldrich

Hafnium

foil, thickness 0.02 mm, 99.5% trace metals basis (purity excludes ~2% zirconium)

Synonym(s):

Celtium, Hafnium element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Hf
CAS Number:
Molecular Weight:
178.49
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

99.5% trace metals basis (purity excludes ~2% zirconium)

form

foil

resistivity

29.6 μΩ-cm, 0°C

thickness

0.02 mm

bp

4602 °C (lit.)

mp

2227 °C (lit.)

density

13.3 g/cm3 (lit.)

SMILES string

[Hf]

InChI

1S/Hf

InChI key

VBJZVLUMGGDVMO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Quantity

3.8 g = 100 × 150 mm

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael J Sgro et al.
Chemical communications (Cambridge, England), 49(26), 2610-2612 (2013-02-07)
Hf-phosphinoamide cation complexes behave as metal-based frustrated Lewis pairs and bind one or two equivalent of CO2 and in as well can activate CO2 in a bimetallic fashion to give a pseudo-tetrahedral P2CO2 fragment linking two Hf centres.
Brian R Dorvel et al.
ACS nano, 6(7), 6150-6164 (2012-06-15)
Nanobiosensors based on silicon nanowire field effect transistors offer advantages of low cost, label-free detection, and potential for massive parallelization. As a result, these sensors have often been suggested as an attractive option for applications in point-of-care (POC) medical diagnostics.
Yu Gong et al.
Dalton transactions (Cambridge, England : 2003), 41(38), 11706-11715 (2012-08-18)
The isolated group 4 metal oxydifluoride molecules OMF(2) (M = Ti, Zr, Hf) with terminal oxo groups are produced specifically on the spontaneous reactions of metal atoms with OF(2) through annealing in solid argon. The product structures and vibrational spectra
Christopher C Underwood et al.
Inorganic chemistry, 52(1), 237-244 (2012-12-19)
This paper describes the hydrothermal chemistry of alkali hafnium fluorides, including the synthesis and structural characterization of five new alkali hafnium fluorides. Two ternary alkali hafnium fluorides are described: Li(2)HfF(6) in space group P31m with a = 4.9748(7) Å and
Suresh Kumar Raman Pillai et al.
ACS applied materials & interfaces, 4(12), 7047-7054 (2012-12-01)
The large-scale application of semiconducting single-walled carbon nanotubes (s-SWCNTs) for printed electronics requires scalable, repeateable, as well as noncontaminating assembly techniques. Previously explored nanotube deposition methods include serial methods such as inkjet printing and parallel methods such as spin-coating with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service