Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

266051

Sigma-Aldrich

Titanium

rod, diam. 6.35 mm, 99.7% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ti
CAS Number:
Molecular Weight:
47.87
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

99.7% trace metals basis

form

rod

autoignition temp.

860 °F

resistivity

42.0 μΩ-cm, 20°C

diam.

6.35 mm

bp

3287 °C (lit.)

mp

1660 °C (lit.)

density

4.5 g/mL at 25 °C (lit.)

SMILES string

[Ti]

InChI

1S/Ti

InChI key

RTAQQCXQSZGOHL-UHFFFAOYSA-N

Quantity

25 g = 16.7 cm; 100 g = 66.8 cm

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Kurbad et al.
International journal of computerized dentistry, 16(2), 125-141 (2013-08-13)
This article presents two novel options for lithium-disilicate restorations supported by single-tooth implants. By using a Ti-Base connector, hybrid abutments and hybrid abutment crowns can be fabricated for different implant systems. The latter option in particular is an interesting new
J H Kim et al.
Journal of nanoscience and nanotechnology, 13(7), 4601-4607 (2013-08-02)
Nanocytalline TiN films were deposited on non-alkali glass and Al substrates by reactive DC magnetron sputtering (DCMS) with an electromagnetic field system (EMF). The microstructure and corrosion resistance of the TiN-coated Al substrates were estimated by X-ray diffraction (XRD), scanning
Jinho Shin et al.
Journal of nanoscience and nanotechnology, 13(8), 5807-5810 (2013-07-26)
In this study, hydroxyapatite (HA) was coated on anodized titanium (Ti) surfaces through radio frequency magnetron sputtering in order to improve biological response of the titanium surface. All the samples were blasted with resorbable blasting media (RBM). RBM-blasted Ti surface
Jiangxue Wang et al.
Journal of nanoscience and nanotechnology, 13(6), 3874-3879 (2013-07-19)
Nanoscale materials (such as TiO2, hydroxyapatite nanoparticles) have gained much concern in the coating of implants for cell adhesion and growth to improve the osteoconductivity. However, due to attrition and corrosion, the wear particles would be generated from the joint
Lu-Ning Wang et al.
Journal of nanoscience and nanotechnology, 13(8), 5316-5326 (2013-07-26)
Electrochemically anodized TiO2 nanotubular arrays can provide large surface areas for biological species attachment. In order to further enhance the biocompatibility of Ti medical implants, we deposited a pre-synthesized hydroxyapatite inside and on the nanotubular arrays, and examined the biocompatibility

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service