Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

26568

Sigma-Aldrich

2-Chlorotrityl chloride, polymer-bound

100-300 mesh, extent of labeling: ~1.6 mmol/g Cl loading

Synonym(s):

2,α-Dichlorobenzhydryl-polystyrene crosslinked with divinylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein/REAXYS Number:
1983914
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

extent of labeling

~1.6 mmol/g Cl loading

matrix

polystyrene, crosslinked with 1% DVB

particle size

100-300 mesh

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

Clc1ccccc1C(Cl)(c2ccccc2)c3ccccc3

InChI

1S/C19H14Cl2/c20-18-14-8-7-13-17(18)19(21,15-9-3-1-4-10-15)16-11-5-2-6-12-16/h1-14H

InChI key

JFLSOKIMYBSASW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Acid labile resin used in Fmoc-based solid phase peptide synthesis.
  • Mild acidic cleavage conditions lead to the release of the peptide acid where fully protected peptides can be released if desired.
  • 2-Chlorotrityl chloride resins prevent racemization of the first amino acid and are thus very useful when racemic mixtures are forming (common with residues such as His or Cys).
  • This resin also prevents diketopiperazide formation, which can be an issue with proline C-terminal peptide sequences.

Use:
  • Attachment of the first amino acid residue is effected by stirring the resin, the protected amino acid, and excess diisopropylethylamine (DIEA) in dichloromethane.
  • Cleavage of the final protected peptide fragment is achieved under very mild conditions using either acetic acid/trifluoroethanol (TFE)/dichloromethane (1:1:8; v/v/v), hexafluoroisopropanol (HFIP)/dichloromethane (1:4; v/v) or simply 0.5% trifluoroacetic acid/dichloromethane (v/v).
  • Higher concentrations of TFA can be used if retention of peptide side chaing protecting groups is unimportant. Note that trityl chloride is moisture-sensitive, and, therefore, should be stored and handled appropriately.
  • If the resin becomes deactivated, treatment with acetyl chloride or SOCl2 in toluene before use is recommended to restore its activity.

Other Notes

Carrier used for solid phase peptide synthesis with FMOC-amino acids.[1][2][3]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liebigs Ann. Chem., 215-215 (1993)
P. Athanassopoulos et al.
Tetrahedron Letters, 36, 5645-5645 (1995)
K. Barlos et al.
Tetrahedron Letters, 30, 3947-3947 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service