Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

265454

Sigma-Aldrich

Cadmium

rod, diam. 4.0 mm, 99.999% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cd
CAS Number:
Molecular Weight:
112.41
Beilstein/REAXYS Number:
8137359
EC Number:
MDL number:
UNSPSC Code:
12141707
PubChem Substance ID:

Quality Level

assay

99.999% trace metals basis

form

rod

resistivity

7.27 μΩ-cm, 22°C

diam.

4.0 mm

bp

765 °C (lit.)

mp

320.9 °C (lit.)

density

8.65 g/mL at 25 °C (lit.)

SMILES string

[Cd]

InChI

1S/Cd

InChI key

BDOSMKKIYDKNTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Quantity

10 g = 100 mm

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2 - Repr. 2 - STOT RE 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Boldizsar Nagy et al.
Acta chimica Slovenica, 60(2), 263-273 (2013-07-25)
Biosorption of cadmium ions from synthetic aqueous solution using popular Romanian fir tree sawdust (Abies Alba) as biosorbent, was investigated in this work. Prior to its utilization the considered biomass was washed, dried and sieved without further chemical treatments. The
Weiping Chen et al.
Environmental pollution (Barking, Essex : 1987), 178, 294-299 (2013-04-18)
Safety of agricultural irrigation with reclaimed water is of great concern as some potential hazardous compounds like heavy metals may be accumulated in soils over time. Impacts of long-term reclaimed water on soil Cd pollution were evaluated based on the
Shigeo Hasegawa et al.
Molecular pain, 5, 22-22 (2009-05-05)
Peripheral nerve injury leads to a persistent neuropathic pain state in which innocuous stimulation elicits pain behavior (tactile allodynia), but the underlying mechanisms have remained largely unknown. We have previously shown that spinal nerve injury induces the activation of cytosolic
F Tomei et al.
Annali di igiene : medicina preventiva e di comunita, 25(4), 353-363 (2013-05-25)
The exposure to Cd has been linked to many disease and liver is the major target organ of Cd toxicity following acute or chronic exposure.No study has ever evaluated the association between liver function tests and exposure to Cd present
Gang Zhang et al.
Journal of environmental sciences (China), 25(1), 231-235 (2013-04-17)
The emission characteristics of polychlorinated dibenzo-p-dioxins and dibenzofurans (PCDD/Fs) and heavy metals were evaluated during co-combustion of sewage sludge with coal from a circulating fluidized bed incinerator. The stack gas, slag and fly ash samples were sampled and analyzed. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service