Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

264997

Sigma-Aldrich

(4S,5S)-2,2-Dimethyl-α,α,α′,α′-tetraphenyldioxolane-4,5-dimethanol

97%

Synonym(s):

(+)-2,3-O-Isopropylidene-1,1,4,4-tetraphenyl-D-threitol, (+)-trans-α,α′-(2,2-Dimethyl-1,3-dioxolane-4,5-diyl)bis(diphenylmethanol), (4S,5S)-4,5-Bis(diphenylhydroxymethyl)-2,2-dimethyldioxolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H30O4
CAS Number:
Molecular Weight:
466.57
Beilstein/REAXYS Number:
4724097
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

97%

optical activity

[α]19/D +67°, c = 1 in chloroform

mp

193-196 °C (lit.)

SMILES string

CC1(C)O[C@@H]([C@H](O1)C(O)(c2ccccc2)c3ccccc3)C(O)(c4ccccc4)c5ccccc5

InChI

1S/C31H30O4/c1-29(2)34-27(30(32,23-15-7-3-8-16-23)24-17-9-4-10-18-24)28(35-29)31(33,25-19-11-5-12-20-25)26-21-13-6-14-22-26/h3-22,27-28,32-33H,1-2H3/t27-,28-/m0/s1

InChI key

OWVIRVJQDVCGQX-NSOVKSMOSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ligand for enantioselective Lewis-acid mediated Diels-Alder,[1] allyltitanation of aldehydes,[2] and Michael-Mukaiyama additions promoted by chiral Ti-complexes.[3]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bernardi, A. et al.
Tetrahedron Letters, 36, 1363-1363 (1995)
Quinkert, G. et al.
Tetrahedron Letters, 33, 3617-3617 (1992)
A. Hafner et al.
Journal of the American Chemical Society, 114, 2321-2321 (1992)

Articles

TADDOL chiral auxiliaries find diverse applications in asymmetric synthesis, catalysis, and polymerization.

TADDOLs demonstrate versatility in metal-catalyzed asymmetric reactions and as Brønsted acid organocatalysts in hetero-Diels–Alder reactions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service