Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

263621

Sigma-Aldrich

Methyl 2,5-dimethyl-3-furancarboxylate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10O3
CAS Number:
Molecular Weight:
154.16
Beilstein/REAXYS Number:
116271
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

refractive index

n20/D 1.475 (lit.)

bp

198 °C (lit.)

density

1.037 g/mL at 25 °C (lit.)

SMILES string

COC(=O)c1cc(C)oc1C

InChI

1S/C8H10O3/c1-5-4-7(6(2)11-5)8(9)10-3/h4H,1-3H3

InChI key

MNFOIGQBJZVPCS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl 2,5-dimethyl-3-furancarboxylate may be used in chemical synthesis.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

177.8 °F - closed cup

flash_point_c

81 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service